Skip to Content
Merck
All Photos(1)

Documents

W359602

Sigma-Aldrich

3,4-Xylenol

≥98%, FG

Synonym(s):

3,4-Dimethylphenol, 4-Hydroxy-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
FEMA Number:
3596
Beilstein:
1099267
EC Number:
Council of Europe no.:
11262
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.048
NACRES:
NA.21

biological source

synthetic

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥98%

bp

227 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

burnt

SMILES string

Cc1ccc(O)cc1C

InChI

1S/C8H10O/c1-6-3-4-8(9)5-7(6)2/h3-5,9H,1-2H3

InChI key

YCOXTKKNXUZSKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Green Aromatic Epoxidation with an Iron Porphyrin Catalyst for One-Pot Functionalization of Renewable Xylene, Quinoline, and Acridine.: This study explores the green epoxidation of aromatic compounds using an iron porphyrin catalyst, demonstrating effective one-pot functionalization of renewable xylene derivatives, including 3,4-xylenol (Corrêa et al., 2023).

  • A highly expressed odorant receptor from the yellow fever mosquito, AaegOR11, responds to (+)- and (-)-fenchone and a phenolic repellent.: This research identifies an odorant receptor in mosquitoes that responds to various compounds, including 3,4-dimethylphenol, which could have implications for developing new repellents (Lu et al., 2022).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A P Hart et al.
Journal of forensic sciences, 42(4), 693-696 (1997-07-01)
Phenol (carbolic acid) is widely used as a disinfectant as well as in the chemical industry as an intermediate in the synthesis of a variety of chemicals. Phenol is also the major metabolite of benzene which is used in many
Maria Concetta Tomei et al.
Environmental science and pollution research international, 15(3), 188-195 (2008-05-28)
In this study, attention was focused on substituted phenols because of their widespread presence in industrial effluents originating from many different sources: they are major constituents of wastewater from coal conversion processes, coke ovens, petroleum refineries and petrochemical industries, resin
M Bartilson et al.
Molecular & general genetics : MGG, 220(2), 294-300 (1990-01-01)
The gene organization of the phenol catabolic pathway of Pseudomonas CF600 has been investigated. This strain can grow on phenol and some methylated phenols by virtue of an inducible phenol hydroxylase and metacleavage pathway enzymes. The genes coding for these
A Dasgupta et al.
Journal of forensic sciences, 42(4), 697-700 (1997-07-01)
Benzyl alcohol is commonly used as an antibacterial agent in a variety of pharmaceutical formulations. Several fatalities in neonates have been linked to benzyl alcohol poisoning. Most methods for measuring benzyl alcohol concentrations in serum utilize direct extraction followed by
H Pavel et al.
Journal of bacteriology, 176(24), 7550-7557 (1994-12-01)
The pVI150 catabolic plasmid of Pseudomonas sp. strain CF600 carries the dmp system, which comprises the divergently transcribed dmpR gene and the dmp operon coding for the catabolic enzymes required for growth on (methyl)phenols. The constitutively expressed DmpR transcriptional activator

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service