Skip to Content
Merck
All Photos(1)

Documents

W330829

Sigma-Aldrich

Methyl propyl trisulfide

50%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10S3
CAS Number:
Molecular Weight:
154.32
FEMA Number:
3308
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.020
NACRES:
NA.21

biological source

synthetic

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

concentration

50%

refractive index

n20/D 1.566

density

1.107 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

garlic; green; metallic; onion; sulfurous

SMILES string

CCCSSSC

InChI

1S/C4H10S3/c1-3-4-6-7-5-2/h3-4H2,1-2H3

InChI key

ZOYASYRPGHCQHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system, Upper respiratory tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Restituto Tocmo et al.
Journal of agricultural and food chemistry, 62(23), 5296-5304 (2014-05-21)
There is a growing account of the health benefits of H2S as an endogenous cell-signaling molecule. H2S from organic polysulfides, in particular, is increasingly gaining attention for their beneficial effects to cardiovascular health. Here, we studied shallot as a potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service