Skip to Content
Merck
All Photos(1)

Key Documents

W325600

Sigma-Aldrich

Benzothiazole

≥96%, FG

Synonym(s):

1-thia-3-azaindene, Vangard BT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NS
CAS Number:
Molecular Weight:
135.19
FEMA Number:
3256
Beilstein:
109468
EC Number:
Council of Europe no.:
11594
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.016
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor density

4.66 (vs air)

vapor pressure

34 mmHg ( 131 °C)

Assay

≥96%

impurities

≤0.1% aniline

refractive index

n20/D 1.642 (lit.)

bp

231 °C (lit.)

mp

2 °C (lit.)

density

1.238 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; meaty; vegetable; brown; nutty; sulfurous

SMILES string

c1ccc2scnc2c1

InChI

1S/C7H5NS/c1-2-4-7-6(3-1)8-5-9-7/h1-5H

InChI key

IOJUPLGTWVMSFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzothiazole, a volatile sulfur-containing heterocyclic compound, is one of the key odorants of heated milk. It is also reported to occur as a flavor component of miso, coconut meat and cooked-beef.

Application


  • Modulating mycobacterial envelope integrity for antibiotic synergy with benzothiazoles.: This publication explores the role of benzothiazoles in enhancing antibiotic efficacy against mycobacteria. The study provides insights into how chemical modification of microbial cell envelopes can improve drug delivery and efficacy (Habjan et al., 2024).

  • An activatable fluorescence probe for rapid detection and in situ imaging of β-galactosidase activity in cabbage roots under heavy metal stress.: This research uses benzothiazole-based probes for real-time and in situ imaging of enzyme activity in plants under stress conditions, facilitating the study of environmental stress impacts on plant physiology and providing a tool for agricultural biotechnology (Zhao et al., 2024).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

224.6 °F - Pensky-Martens closed cup

Flash Point(C)

107 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal generation of sulfur-containing flavor compounds in beef.
Vercellotti JR, et al.
ACS Symp. Ser., 409, 452?459-452?459 (1989)
Gas chromatography olfactometry (GC/O) of dairy products.
Friedrich JE & Acree TE.
International dairy journal, 8(3), 235-241 (1998)
Flavor components of miso: basic fraction.
Mori Y, et al.
Agricultural and Biological Chemistry, 47(7), 1487-1492 (1983)
Britt-Marie Swahn et al.
Bioorganic & medicinal chemistry letters, 22(13), 4332-4337 (2012-06-01)
The synthesis and SAR of new β-amyloid binding agents are reported. Evaluation of important properties for achieving good signal-to-background ratio is described. Compounds 27, 33, and 36 displayed desirable lipophilic and pharmacokinetic properties. Compound 27 was further evaluated with autoradiographic
Thanh Binh Nguyen et al.
Angewandte Chemie (International ed. in English), 53(50), 13808-13812 (2014-10-10)
The disproportionation of elemental sulfur at moderate temperatures is investigated in the redox condensation involving o-halonitrobenzenes 1 and benzylamines 2. As a redox moderator, elemental sulfur plays the dual role of both electron donor and acceptor, generating its lowest and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service