Skip to Content
Merck
All Photos(1)

Documents

W321508

Sigma-Aldrich

trans-2-Octenal

≥95%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCHO
CAS Number:
Molecular Weight:
126.20
FEMA Number:
3215
EC Number:
Council of Europe no.:
663
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.190
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

vapor density

>1 (vs air)

Assay

≥95%

contains

α-tocopherol as stabilizer (synthetic)

refractive index

n20/D 1.45 (lit.)

bp

84-86 °C/19 mmHg (lit.)

density

0.846 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; green; herbaceous

SMILES string

[H]C(=O)C(\[H])=C(/[H])CCCCC

InChI

1S/C8H14O/c1-2-3-4-5-6-7-8-9/h6-8H,2-5H2,1H3/b7-6+

InChI key

LVBXEMGDVWVTGY-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-2-octenal is a volatile organic compound that has a nutty and fatty odor. It can be used as a flavoring agent in the food and cosmetic industries.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tolgahan Kocadağlı et al.
Food & function, 3(9), 970-975 (2012-07-17)
Coffee, as a source of acrylamide, needs to be investigated in depth to understand the contribution of different precursors. This study aimed to investigate the contributions of sucrose decomposition and lipid oxidation on acrylamide formation in coffee during roasting. Coffee
Chemical composition and antioxidant and antibacterial activities of an essential oil extracted from an edible seaweed, Laminaria japonica L.
P Jayanta Kumar, et al.
Molecules (Basel), 20(7), 12093-12113 (2015)
Yin Zhu et al.
Food research international (Ottawa, Ont.), 108, 74-82 (2018-05-08)
A chestnut-like aroma is widely considered an important indicator of an excellent-quality green tea; however, the key odorants responsible for chestnut-like aroma have never been systematically studied and remain unknown. In this study, the aroma components of green teas and
Michaela Bodner et al.
Journal of chemical ecology, 38(5), 547-556 (2012-05-15)
The secretions from serial defensive glands of the Austrian diplopod Allajulus dicentrus (Julidae, Cylindroiulini) were extracted and analyzed by means of gas chromatography - mass spectrometry. In adults, 13 components from two chemical classes were detected: 1) The common juliform
K Hasegawa et al.
Biochimica et biophysica acta, 962(3), 371-376 (1988-10-14)
The structures of fluorescent products formed in the reaction of methyl linoleate hydroperoxides with adenine, FeSO4 and ascorbic acid were investigated to elucidate the mechanism of interaction. The fluorescent products consisted of at least four major components (I-IV), which could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service