Skip to Content
Merck
All Photos(1)

Key Documents

W258806

Sigma-Aldrich

4-(4-Hydroxyphenyl)-2-butanone

≥98%, FCC, FG

Synonym(s):

Frambione, NSC 26515, Raspberry ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2COCH3
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2588
Beilstein:
776080
EC Number:
Council of Europe no.:
755
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.055
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

mp

81-85 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

berry; jam; raspberry; sweet

SMILES string

CC(=O)CCc1ccc(O)cc1

InChI

1S/C10H12O2/c1-8(11)2-3-9-4-6-10(12)7-5-9/h4-7,12H,2-3H2,1H3

InChI key

NJGBTKGETPDVIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Böker et al.
Biotechnology progress, 17(3), 568-572 (2001-06-02)
The basidiomycete Nidula niveo-tomentosa produced 4-(4-hydroxyphenyl)-butan-2-one (raspberry ketone), one of the character impact components of raspberry flavor, and its corresponding alcohol. A systematic attempt was made to improve the productivity of this fungus. Variation of nutrient medium composition, precursor amount
Naoaki Harada et al.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society, 18(4), 335-344 (2008-03-07)
Sensory neurons release calcitonin gene-related peptide (CGRP) on activation. We recently reported that topical application of capsaicin increases facial skin elasticity and promotes hair growth by increasing dermal insulin-like growth factor-I (IGF-I) production through activation of sensory neurons in mice
H Zorn et al.
Applied and environmental microbiology, 69(1), 367-372 (2003-01-07)
Submerged cells of the basidiomycete Nidula niveo-tomentosa, a microbial producer of 4-(4-hydroxyphenyl)-butan-2-one, were supplemented with (13)C-labeled L-phenylalanines and with [1-(13)C]glucose. Labeled transformation products were detected by a novel method of analyzing stable isotope-labeled metabolites, gas chromatography (GC) coupled to an
S Pedapudi et al.
Biotechnology progress, 16(3), 346-349 (2000-06-03)
Production levels of p-coumaric acid (p-CA), p-hydroxyphenylbut-3-ene-2-one (benzalacetone), and p-hydroxyphenyl-2-butanone (raspberry ketone) were measured in raspberry cell suspension cultures to investigate metabolite dynamics in a short (two-step) pathway. Intracellular concentrations of benzalacetone and the raspberry ketone fluctuated during the time
Kyoung Sik Park
Planta medica, 76(15), 1654-1658 (2010-04-29)
Raspberry ketone (RK) is a natural phenolic compound of the red raspberry. The dietary administration of RK to male mice has been reported to prevent high-fat diet-induced elevation in body weight and to increase lipolysis in white adipocytes. To elucidate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service