Skip to Content
Merck
All Photos(2)

Key Documents

T13005

Sigma-Aldrich

1,2,3,4-Tetrahydroisoquinoline

95%

Synonym(s):

1,2,3,4-Tetrahydro-2-azanaphthalene, 1,2,3,4-Tetrahydro-2-isoquinoline, 1,2,3,4-Tetrahydroisoquinline, 1,2,3,4-Tetrahydroisoquinoline, 3,4-Dihydro-1H-isoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
Beilstein:
116156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

refractive index

n20/D 1.568 (lit.)

bp

232-233 °C (lit.)

mp

−30 °C (lit.)

density

1.064 g/mL at 25 °C (lit.)

SMILES string

C1Cc2ccccc2CN1

InChI

1S/C9H11N/c1-2-4-9-7-10-6-5-8(9)3-1/h1-4,10H,5-7H2

InChI key

UWYZHKAOTLEWKK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary L Grunewald et al.
Journal of medicinal chemistry, 49(18), 5424-5433 (2006-09-01)
3-Fluoromethyl-7-(N-substituted aminosulfonyl)-1,2,3,4-tetrahydroisoquinolines (14, 16, and 18-22) are highly potent and selective inhibitors of phenylethanolamine N-methyltransferase (PNMT). Molecular modeling studies with 3-fluoromethyl-7-(N-alkyl aminosulfonyl)-1,2,3,4-tetrahydroisoquinolines, such as 16, suggested that the sulfonamide -NH- could form a hydrogen bond with the side chain of
Lucie Cahlíková et al.
Natural product communications, 5(7), 1035-1038 (2010-08-26)
The roots and aerial parts of Eschscholzia californica Cham. (Papaveraceae) were extracted with ethanol. Repeated column chromatography, preparative TLC, and crystallization led to the isolation of fourteen isoquinoline alkaloids, the structures of which were determined on the basis of spectroscopic
Ajay Kumar Srivastava et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(17), 4905-4913 (2011-03-16)
We herein report a robust and efficient synthetic route to highly functionalized enantiopure 1,2,3,4-tetrahydroisoquinolines (THIQs) from Garner aldehyde. We utilized the inherent chirality of Garner aldehyde through 1,2- and 1,3-/1,4-asymmetric inductions iteratively to obtain 1,2,3,4-tetrasubstitued THIQs using rigid and isolable
Matthias Ludwig et al.
European journal of medicinal chemistry, 41(8), 1003-1010 (2006-05-06)
A series of 1-aryl-1,2,3,4-tetrahydroisoquinoline and 8-methyl-1-aryl-1,2,3,4-tetrahydroisoquinoline derivatives was evaluated for affinity to the PCP binding site of the NMDA receptor complex. The (S)-configured tetrahydroisoquinoline derivative (S)-4 e x HCl bearing a 2-methylphenyl substituent in position 1 of the heterocyclic ring
Staffan Skogvall et al.
Pulmonary pharmacology & therapeutics, 21(1), 125-133 (2007-03-22)
Current drugs including beta-agonists have limited smooth muscle relaxant effects on human small airways. Yet this is a major site of obstruction in asthma and chronic obstructive pulmonary disease (COPD). This study explores human small airway relaxant effects of RESPIR

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service