Skip to Content
Merck
All Photos(1)

Documents

SYX00317

Sigma-Aldrich

2-(N-Anilino)pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N3
CAS Number:
Molecular Weight:
171.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

C1(NC2=CC=CC=C2)=NC=CC=N1

InChI

1S/C10H9N3/c1-2-5-9(6-3-1)13-10-11-7-4-8-12-10/h1-8H,(H,11,12,13)

InChI key

XGXNTJHZPBRBHJ-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Bernabò et al.
Aquatic toxicology (Amsterdam, Netherlands), 172, 56-66 (2016-01-16)
Over the last few years, the hazards associated with the extensive use of fungicides have become an issue of great concern but, at present, the effects of these substances on amphibians remain poorly understood. The goal of the present study
F A Esteve-Turrillas et al.
The Analyst, 142(20), 3975-3985 (2017-09-29)
The development of multianalyte immunoassays constitutes a main research issue in the field of bioanalytical techniques. In the present study, class-specific antibodies against the three members of the anilinopyrimidine family of fungicides (pyrimethanil, cyprodinil and mepanipyrim) were raised by using
Jeffery A DeLong et al.
Phytopathology, 110(3), 694-702 (2020-02-06)
Botrytis cinerea, the causal agent of gray mold, has high genetic diversity and a broad host range. In Vitis sp. and Prunus spp., B. cinerea causes pre- and postharvest diseases, and fungicides are routinely applied to prevent yield loss. In
Marcos Seoane et al.
Oncogene, 38(19), 3616-3635 (2019-01-18)
The melanocytic lineage, which is prominently exposed to ultraviolet radiation (UVR) and radiation-independent oxidative damage, requires specific DNA-damage response mechanisms to maintain genomic and transcriptional homeostasis. The coordinate lineage-specific regulation of intricately intertwined DNA repair and transcription is incompletely understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service