Key Documents
SYNC101100
Synthware Vigreux distillation column
joint: ST/NS 14/20, L 100 mm
Synonym(s):
Synthware C101100, distilling column, fractional column
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C101100
L
100 mm
General description
Column length is measured from the highest to lowest indent in the column.
Legal Information
Synthware is a trademark of Kemtech America Inc.