SLW4526/25D
Pyrex® High temperature screwcap for Media-lab bottle
red, thread suits 25 mm
Synonym(s):
Pyrex, SciLabware 4526/25D
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
PTFE liner
polyester cap
red
manufacturer/tradename
4526/25D
SciLabware
fitting
thread suits 25 mm
Looking for similar products? Visit Product Comparison Guide
General description
- For use with 1516, 1517, 1518, 1519 and 1520 series bottles
- Ideal for applications involving high temperatures or aggressive chemicals
- Red thermoplastic polyester cap with natural coloured PTFE faced sealing disc
- Withstands hot air sterilisation up to 180 deg. C
- PTFE sealing disc resists chemical attack
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service