Skip to Content
Merck
All Photos(1)

Key Documents

S5502

Sigma-Aldrich

Succinic dihydrazide

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NNHCOCH2CH2CONHNH2
CAS Number:
Molecular Weight:
146.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

mp

170-171 °C (lit.)

SMILES string

NNC(=O)CCC(=O)NN

InChI

1S/C4H10N4O2/c5-7-3(9)1-2-4(10)8-6/h1-2,5-6H2,(H,7,9)(H,8,10)

InChI key

HCOMFAYPHBFMKU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena A Khomyakova et al.
Nucleosides, nucleotides & nucleic acids, 30(7-8), 577-584 (2011-09-06)
An efficient method for the synthesis of DNA or RNA oligonucleotide 2'-hydrazides is described. Fully deprotected oligonucleotides containing a hydrazide group at the 2'-position of a uridine residue were obtained by a novel two-step procedure: periodate cleavage of an oligonucleotide
Marlene Lopes et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 113, 88-96 (2016-12-23)
This study aimed to assess the biodistribution of antihyperglycemic insulin-loaded alginate/dextran sulfate-based nanoparticles dual coated with chitosan and technetium-99m-albumin (
N Yoshitani et al.
Analytical biochemistry, 277(1), 127-134 (1999-12-28)
We developed a convenient method for the synthesis of dextran-based multivalent probes containing N-linked oligosaccharides which is efficient even in a small scale. Oligosaccharides were derivatized with succinic dihydrazide and dimethylamine borane under a mild acidic condition. The derivatized oligosaccharides
Leeanne McGurk et al.
Molecular cell, 71(5), 703-717 (2018-08-14)
In amyotrophic lateral sclerosis (ALS) and frontotemporal degeneration (FTD), cytoplasmic aggregates of hyperphosphorylated TDP-43 accumulate and colocalize with some stress granule components, but how pathological TDP-43 aggregation is nucleated remains unknown. In Drosophila, we establish that downregulation of tankyrase, a poly(ADP-ribose)
Yerang Seo et al.
Carbohydrate polymers, 218, 1-7 (2019-06-22)
Tissue engineering typically requires a use of scaffolds when delivering tissue-specific cells to be engineered. Hydrogels are frequently used as scaffolds, because their composition, structure, and function resemble the natural tissue extracellular matrix. In this study, hyaluronate-alginate hybrid (HAH) was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service