Skip to Content
Merck
All Photos(4)

Key Documents

S4007

Sigma-Aldrich

Sodium iodate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaIO3
CAS Number:
Molecular Weight:
197.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes

grade

for analytical purposes

Assay

≥99%

density

4.28 g/mL at 25 °C (lit.)

SMILES string

[Na+].[O-]I(=O)=O

InChI

1S/HIO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

WTCBONOLBHEDIL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium iodate (NaIO3) can be used as an oxidant in the following processes:     
  • NaIO3/neutral Al2O3 catalytic system is used to synthesize a variety of aromatic, aliphatic, and alicyclic disulfides via oxidation of the corresponding thiols.      
  • NaIO3/Na2SO3/HCl is used in the preparation of 2-amino-1-iodonaphthalene by iodination of 2-naphthylamine.
  • NaIO3/acetic anhydride or glacial acetic acid is used in aromatic oxidative iodination reactions.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodination of both deactivated and activated arenes with sodium periodate or sodium iodate as the oxidants
Lulinski P and Skulski L
Bulletin of the Chemical Society of Japan, 73(4), 951-956 (2000)
Simple Oxidation of Thiols with Sodium Iodate under Solid-Solution Biphasic Conditions
Hirano M, et al.
Journal of chemical research. Synopses, (12), 816-817 (1998)
Jie Huang et al.
Molecular vision, 26, 494-504 (2020-07-09)
To observe the effects of tetramethylpyrazine (TMP) on mice retinas injured by sodium iodate (NaIO3). Male mice (n = 45) were randomly divided into three groups: the control group (Group C), the NaIO3-degenerated group (Group I), and the TMP-treated group
Reaction of 2-Naphthylamine with Sodium Iodate/Sodium Sulfite: Synthesis of 2-Amino-1-iodonaphthalene
Taldone T, et al.
Synthetic Communications, 42(5), 635-638 (2012)
Brianna M Young et al.
Molecular therapy. Methods & clinical development, 14, 113-125 (2019-07-25)
Age-related macular degeneration (AMD) has been linked to oxidative damage and para-inflammation, an activation of inflammasome signaling in the retinal pigment epithelium (RPE) and the underlying choriocapillaris. Herein, we tested the efficacy of a gene-delivered caspase-1 inhibitor in controlling the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service