Skip to Content
Merck
All Photos(1)

Documents

P66009

Sigma-Aldrich

3-Pyridylacetonitrile

98%

Synonym(s):

3-Pyridineacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2
CAS Number:
Molecular Weight:
118.14
Beilstein:
111292
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.529 (lit.)

bp

101-109 °C/1.5 mmHg (lit.)

density

1.108 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

N#CCc1cccnc1

InChI

1S/C7H6N2/c8-4-3-7-2-1-5-9-6-7/h1-2,5-6H,3H2

InChI key

OIPHWUPMXHQWLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Andharia et al.
Pflugers Archiv : European journal of physiology, 470(6), 897-907 (2018-02-06)
The pancreatic duct epithelium secretes the HCO3--rich pancreatic juice. The HCO3- transport across the luminal membrane has been proposed to be mediated by SLC26A Cl--HCO3- exchangers. To examine the electrophysiological properties of Cl--HCO3- exchangers, we directly measured HCO3- conductance in
Magalí Pasqualone et al.
Materials science & engineering. C, Materials for biological applications, 76, 652-658 (2017-05-10)
Transdermal drug delivery system (TDDS) could be seen as alternative to the oral administration which avoids several adverse effects. In this study a novel TDDS for risedronate (RI), a bisphosphonate used for osteoporosis treatment, based on a vinyl acetate-dioctylfumarate copolymer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service