Skip to Content
Merck
All Photos(2)

Documents

O4709

Sigma-Aldrich

2-Octanone

reagent grade, 98%

Synonym(s):

Hexyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COCH3
CAS Number:
Molecular Weight:
128.21
Beilstein:
635843
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Assay

98%

refractive index

n20/D 1.416 (lit.)

bp

173 °C (lit.)

mp

−16 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(C)=O

InChI

1S/C8H16O/c1-3-4-5-6-7-8(2)9/h3-7H2,1-2H3

InChI key

ZPVFWPFBNIEHGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J S Hosler et al.
The Journal of experimental biology, 203(Pt 18), 2797-2806 (2000-08-23)
Recent studies of olfactory blocking have revealed that binary odorant mixtures are not always processed as though they give rise to mixture-unique configural properties. When animals are conditioned to one odorant (A) and then conditioned to a mixture of that
Jian Hu et al.
Biotechnology letters, 28(14), 1115-1119 (2006-06-24)
An aqueous-organic biphasic system was established and used with whole cells of Oenococcus oeni to reduce 2-octanone to (R)-2-octanol. The conversion reached 99% when the Tris/borate buffer was increased from 50 mM to 300 mM in the aqueous phase. In
M Krogh et al.
Journal of chromatography. B, Biomedical sciences and applications, 689(2), 357-364 (1997-02-21)
This paper describes microextraction and gas chromatographic analysis of diazepam from human plasma. The method was based on immobilisation of 1.5 microliters of 1-octanol on a polyacrylate-coated fiber designed for solid-phase microextraction. The solvent-modified fibre was used to extract diazepam
J Misumi et al.
Sangyo igaku. Japanese journal of industrial health, 24(5), 475-484 (1982-09-01)
An electrophysiological study of the neurotoxicity of 2-octanone (an analogue of methyl n-butyl ketone) and 2-hexanol (a metabolite of n-hexane) was conducted on rats as a part of the study to determine the specific molecular arrangement required for the development
D G Laing et al.
Chemical senses, 28(1), 57-69 (2002-12-28)
Increasing the concentration of an odorant increases the number of receptor cells and glomeruli in the olfactory bulb that are stimulated, and it is commonly acknowledged that these represent increased numbers of receptor types. Currently, it is not known whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service