Skip to Content
Merck
All Photos(2)

Key Documents

O3132

Sigma-Aldrich

Oxibendazole

~98%

Synonym(s):

Methyl (5-propoxy-1H-benzimidazol-2-yl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O3
CAS Number:
Molecular Weight:
249.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

~98%

Quality Level

storage temp.

2-8°C

SMILES string

CCCOc1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O3/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

RAOCRURYZCVHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anthelminthic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G W Lubega et al.
Biochemical pharmacology, 41(1), 93-101 (1991-01-01)
The low- and high-affinity binding of tritiated benzimidazole anthelmintics (mebendazole and oxibendazole) to tubulin-containing supernatants derived from unembryonated eggs, third stage larvae or adult worms of thiabendazole-susceptible and -resistant strains of Haemonchus contortus were examined and compared. The displacement of
Ray M Kaplan et al.
Journal of the American Veterinary Medical Association, 225(6), 903-910 (2004-10-16)
To determine prevalence of anthelmintic resistance in cyathostome nematodes of horses in the southern United States. Cross-sectional study. 786 horses on 44 farms and stables in Georgia, South Carolina, Florida, Kentucky, and Louisiana. Fecal egg count (FEC) reduction tests were
F F Franssen et al.
Antimicrobial agents and chemotherapy, 39(6), 1265-1268 (1995-06-01)
In the light of the increased incidence of human Encephalitozoon infections and the absence of an established treatment protocol, a simple in vitro testing method to compare activities of drugs against Encephalitozoon cuniculi was developed. With this in vitro method
C Gokbulut et al.
Research in veterinary science, 72(1), 11-15 (2002-05-11)
Oxibendazole (OBZ) was administered to eight horses at an oral dose of 10 mg kg(-1) bodyweight each. Parent OBZ could only be detected in plasma at the 0.5 and 1.0 hours post administration sampling times and the mean maximum plasma
R P Herd et al.
Veterinary parasitology, 48(1-4), 229-240 (1993-06-01)
Environmental consequences of treating horses with recommended dosages of ivermectin paste were studied in two controlled experiments with 29 horses in Ohio. In 1988, dung dispersal rates were measured by changes in dry weight over time of 48 copromes (300

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service