All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99%
bp
303 °C/725 mmHg (lit.)
mp
73-76 °C (lit.)
SMILES string
Cc1ccc2OC(=O)C=Cc2c1
InChI
1S/C10H8O2/c1-7-2-4-9-8(6-7)3-5-10(11)12-9/h2-6H,1H3
InChI key
FXFYOPQLGGEACP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Barry C Pemberton et al.
Chemical communications (Cambridge, England), 46(2), 225-227 (2009-12-22)
Cucurbit[8]uril (as low as 10 mol%) acts as a supramolecular catalytic nanoreaction vessel and facilitates the photodimerization of coumarins in water leading to syn dimers. Saturation kinetics shows a sigmoidal dependence with a turnover number of 3.4 min(-1) and a
Veenasangeeta Sortur et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(2), 301-307 (2006-04-29)
Fourier transform-infrared (4000-400 cm-1) and Raman (3500-50 cm-1) spectral measurements have been made for 6-methyl-4-bromomethylcoumarin. Equilibrium structures, harmonic vibrational frequencies, infrared intensities, and depolarization ratios have been computed at RHF/6-31G* and B3LYP/6-31G* levels of theory. Twisting CH2Br moiety in the
Feiying Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(2), 710-713 (2008-03-25)
The infrared absorption spectra of the carbonyl stretching vibrations of 6-methylcoumarin (6MC) have been investigated in CCl4/ROH mixtures (CCl4/C2H5OH, CCl4/n-C3H7OH, CCl4/i-C3H7OH, and CCl4/t-C5H11OH). Two types of carbonyl stretching vibration bands for 6MC are found with the change of the mole
Masatoshi Hoya et al.
Toxicology in vitro : an international journal published in association with BIBRA, 23(5), 911-918 (2009-04-11)
In this study, with the aim of developing a cell-based in vitro photosensitization assay, we examined whether changes of CD86 and CD54 expression on cells of a human monocytic cell line, THP-1, could be used to assess the photosensitizing potential
W P Jordan
Contact dermatitis, 8(2), 109-116 (1982-03-01)
This report describes modifications in the techniques used for both the induction and elicitation of photoallergic contact dermatitis (PACD) in the guinea pig. These changes have improved the reliability of this animal as the model of choice for screening chemicals
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service