Skip to Content
Merck
All Photos(1)

Documents

M31503

Sigma-Aldrich

α-Methylbenzyl cyanide

96%

Synonym(s):

α-Methylphenylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)CN
CAS Number:
Molecular Weight:
131.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.5106 (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

CC(C#N)c1ccccc1

InChI

1S/C9H9N/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8H,1H3

InChI key

NVAOLENBKNECGF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamás Plaszkó et al.
Metabolites, 10(11) (2020-11-12)
The interaction between plant defensive metabolites and different plant-associated fungal species is of high interest to many disciplines. Volatile organic compounds (VOCs) are natural products that are easily evaporated under ambient conditions. They play a very important role in inter-species
Zhong-Liu Wu et al.
Biotechnology and applied biochemistry, 35(Pt 1), 61-67 (2002-02-09)
Racemic 2-phenylpropionitrile was resolved enantioselectively by nitrile-converting enzymes in cells of Rhodococcus sp. CGMCC 0497 to S-(+)-2-phenylpropionic acid and R-(-)-2-phenylpropionamide. By optimization of the culture conditions, great enhancement of enzyme activity and enantioselectivity was achieved. Furthermore, the relationship between cell-growth
Ondřej Kaplan et al.
BMC biotechnology, 11, 2-2 (2011-01-08)
Nitrilases attract increasing attention due to their utility in the mild hydrolysis of nitriles. According to activity and gene screening, filamentous fungi are a rich source of nitrilases distinct in evolution from their widely examined bacterial counterparts. However, fungal nitrilases
Zsolt Szűcs et al.
BMC plant biology, 18(1), 85-85 (2018-05-11)
The health of plants is heavily influenced by the intensively researched plant microbiome. The microbiome has to cope with the plant's defensive secondary metabolites to survive and develop, but studies that describe this interaction are rare. In the current study
A Chmura et al.
Applied microbiology and biotechnology, 81(2), 371-378 (2008-09-17)
An enrichment culture from saline soda soils, using acetate as carbon and energy source and 2-phenylpropionitrile as nitrogen source (PPN) at pH 10, resulted in the isolation of strain ANL-alpha CH3. The strain was identified as a representative of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service