Skip to Content
Merck
All Photos(1)

Documents

M12207

Sigma-Aldrich

3-Methoxy-1-butanol

99%

Synonym(s):

1,3-Butylene glycol 3-monomethyl ether, 3-Methoxybutanol, 3-Methoxybutyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OCH3)CH2CH2OH
CAS Number:
Molecular Weight:
104.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.416 (lit.)

density

0.928 g/mL at 25 °C (lit.)

SMILES string

COC(C)CCO

InChI

1S/C5H12O2/c1-5(7-2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

JSGVZVOGOQILFM-UHFFFAOYSA-N

Application

3-Methoxy-1-butanol can be used as:
  • A solvent in the synthesis of potassium 3-methoxy-1-butylxanthate, an intermediate used in the preparation of xanthate complexes.
  • A model compound in the study of dehydration of alcohols using CeO2 as a catalyst.
  • An intermediate to synthesize perylene diimide-based dyes applicable as the colorant of the black matrix.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydration of diols catalyzed by CeO2
Sato S, et al.
J. Mol. Catal. A: Chem., 221(1-2), 177-183 (2004)
Novel xanthate complexes for the size-controlled synthesis of copper sulfide nanorods
Al-Shakban M, et al.
Inorganic Chemistry, 56(15), 9247-9254 (2017)
Synthesis of bay-linked perylene dimers with enhanced solubility for high optical density black matrix material
Yuk SB, et al.
Dyes and Pigments, 171, 107695-107695 (2019)
Yanqin Gao et al.
The Journal of chemical physics, 142(21), 214505-214505 (2015-06-08)
The dielectric relaxations in six primary and secondary alkoxy alcohols with varying molecular size and different separation between -O- and hydroxyl group are studied at temperatures around glass transition. The analyses of the apparent full width at half maximum of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service