LGFN30K
Near-IR Fluorescent PLGA nanoparticles
30 μm average diameter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
description
Excitation/Emission: 780/820 nm
Excitation/Emission: 460/500 nm
form
pellets
average diameter
30 μm
storage temp.
-10 to -25°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Particles made of poly(D,L-lactide-co-glycolide) L/G ratio of 50/50, inherent viscosity of ~0.45 dL/g.To re-constitute:
- Warm to ambient temperature
- Add DI water or buffer to achieve desired concentration
- Vortex and sonicate for 5 minutes to ensure particles are well suspended
Application
Fluorescent PLGA particles for imaging and cell tracking in drug discovery. Particles can be used for fluorscent assays and drug delivery.
Legal Information
Product of Phosphorex, Inc.Degradex(RM) is a product of Phosphorex, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service