Skip to Content
Merck
All Photos(1)

Key Documents

LG20K

Sigma-Aldrich

PLGA microspheres

20 μm average diameter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

pellets

average diameter

20 μm

Looking for similar products? Visit Product Comparison Guide

General description

Particles made of poly(D,L-lactide-co-glycolide) L/G ratio of 50/50, inherent viscosity of ~0.45 dL/gTo re-constitute:
  • Warm to ambient temperature
  • Add DI water or buffer to achieve desired concentration
  • Vortex and sonicate for 5 minutes to ensure particles are well suspended

Application

Plain PLGA particles for testing system compatibility in drug delivery research. Particles can be used as carriers by attaching drug or biological/chemical entities to particle surface. PLGA particles are used for controlled release, sustained release and targeted drug delivery.

Legal Information

Product of Phosphorex, Inc.Degradex(RM) is a product of Phosphorex, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service