Skip to Content
Merck
All Photos(1)

Key Documents

H5308

Sigma-Aldrich

Hexachloro-2-propanone

99%

Synonym(s):

Hexachloroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3COCCl3
CAS Number:
Molecular Weight:
264.75
Beilstein:
1707475
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.511 (lit.)

bp

66-70 °C/6 mmHg (lit.)

mp

−6-−2 °C (lit.)

density

1.743 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)C(=O)C(Cl)(Cl)Cl

InChI

1S/C3Cl6O/c4-2(5,6)1(10)3(7,8)9

InChI key

DOJXGHGHTWFZHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used for the conversion of nucleoside-3′-phosphonates to the corresponding phosphates and for the polymerization of vinyl compounds.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Macromolecules, 26, 7089-7089 (1993)
H Zochlinski et al.
Mutation research, 89(2), 137-144 (1981-06-01)
Hexachloroacetone, CCl3--CO--CCl3, reverts the Ames strains TA98 and TA100 but not the non-plasmid strains TA1537, TA1535 and TA1538. In the absence of solvent, the number of revertant colonies is 5 times the spontaneous reversion rate for TA100 and 10 times
Determination of hexachloroacetone in air.
E Kissa
Analytical chemistry, 55(8), 1222-1225 (1983-07-01)
Journal of the Chemical Society. Perkin Transactions 1, 1269-1269 (1987)
E R Nestmann et al.
Environmental mutagenesis, 7(2), 163-170 (1985-01-01)
Reports in the literature describing artifactual results owing to interactions of test materials with solvents are becoming more frequent. The present study was initiated to examine possible interactions of 1,1,3-trichloro-, 1,1,3,3-tetrachloro-, pentachloro- and hexachloroacetones with different solvents, since certain solvent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service