The solubility of this product in DMSO has not been determined. However, various sources state that the product can be in DMSO at 1 mg/ml. Please note that this information has not been validated internally,
About This Item
Recommended Products
Quality Level
Assay
≥98%
mp
171-172 °C (lit.)
SMILES string
OC1=C(Oc2ccccc2C1=O)c3ccccc3
InChI
1S/C15H10O3/c16-13-11-8-4-5-9-12(11)18-15(14(13)17)10-6-2-1-3-7-10/h1-9,17H
InChI key
HVQAJTFOCKOKIN-UHFFFAOYSA-N
Gene Information
mouse ... Hexa(15211)
Looking for similar products? Visit Product Comparison Guide
Application
- Studies of photochemically-induced dioxygenase-type CO-release reactivity
- Phase-transfer protection and deprotection of hydroxychromones
- O-methylation with di-Me carbonate
Reactant involved in the synthesis of biologically active molecules including:
- 2-Chloropyridine derivatives for studies of antitumor agents and telomerase inhibitors
- Dihydrochromenopyrazines and chromenoquinoxalines
Involved in studies of its electrochemical properties using voltammetric methodologies
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the solubility of 3-hydroxyflavone in DMSO?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service