Skip to Content
Merck
All Photos(2)

Key Documents

H15403

Sigma-Aldrich

L-Histidine methyl ester dihydrochloride

97%

Synonym(s):

(S)-Histidine methyl ester dihydrochloride, Methyl L-histidinate dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3O2 · 2HCl
CAS Number:
Molecular Weight:
242.10
Beilstein:
3572009
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +9.0°, c = 2 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

207 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Cl.Cl.COC(=O)[C@@H](N)Cc1c[nH]cn1

InChI

1S/C7H11N3O2.2ClH/c1-12-7(11)6(8)2-5-3-9-4-10-5;;/h3-4,6H,2,8H2,1H3,(H,9,10);2*1H/t6-;;/m0../s1

InChI key

DWAYENIPKPKKMV-ILKKLZGPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Histidine methyl ester dihydrochloride can be used as a reactant to synthesize:
  • Imidazopyridine derivatives by Pictet-Spegler reaction with different aldehydes.
  • A metal-chelating ligand, N-methacryloyl-(l)-histidine methyl ester by reacting with methacryloyl chloride.
  • Zwitterionic polypeptide derivative by amidation reaction with poly (α,β-L-aspartic acid).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Scott Worley et al.
Proteins, 46(3), 321-329 (2002-02-09)
Histidine decarboxylase (HDC) from Lactobacillus 30a converts histidine to histamine, a process that enables the bacteria to maintain the optimum pH range for cell growth. HDC is regulated by pH; it is active at low pH and inactive at neutral
Borislav Kovacević et al.
The journal of physical chemistry. A, 109(37), 8329-8335 (2006-07-13)
Gas-phase H/D exchange experiments with CD3OD and D2O and quantum chemical ab initio G3(MP2) calculations were carried out on protonated histidine and protonated histidine methyl ester in order to elucidate their bonding and structure. The H/D exchange experiments show that
Xiaoyu Su et al.
Chirality, 21(5), 539-546 (2008-08-14)
Two kinds of novel chiral molecular tweezers containing imidazoliums were synthesized from L-alanine, L-phenylalanine, and L-glutamic acid. They are constructed by the chiral imidazolium pincers and two different spacers which are 1,3-bis (bromomethyl)benzene and 2,6-bis(bromomethyl)pyridine, respectively. The enantioselective recognition of
Novel metal-chelate affinity sorbents for reversible use in catalase adsorption
Akgol S and Denizli A
Journal of Molecular Catalysis. B, Enzymatic, 28(1), 7-14 (2004)
T A Alston et al.
Biochemistry, 26(13), 4082-4085 (1987-06-30)
L-Histidine methyl ester inactivates histidine decarboxylase in a time-dependent manner. The possibility was considered that an irreversible reaction between enzyme and inhibitor occurs [Recsei, P. A., & Snell, E. E. (1970) Biochemistry 9, 1492-1497]. We have confirmed time-dependent inactivation by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service