GF94607040
Titanium
tube, 1000mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Synonym(s):
Titanium, TI007250
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
≥99.6%
form
tubes
autoignition temp.
860 °F
manufacturer/tradename
Goodfellow 946-070-40
resistivity
42.0 μΩ-cm, 20°C
bp
3287 °C (lit.)
mp
1660 °C (lit.)
density
4.5 g/mL at 25 °C (lit.)
SMILES string
[Ti]
InChI
1S/Ti
InChI key
RTAQQCXQSZGOHL-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of nanoscience and nanotechnology, 14(6), 4387-4393 (2014-04-18)
The TiO2 nanotubes by anodization have been extensively studied for medical implant and orthopedic applications because of enhancing bone development. In the present study, a new nano-foveolae structure verified by SEM and AFM was prepared by simulating the nanotubes exfoliation
Journal of nanoscience and nanotechnology, 14(6), 4292-4296 (2014-04-18)
The structural and functional characterizations of titanium nitride (TiN) advanced ceramic microsprings (CMSs), with a coil diameter of several micrometers and synthesized by chemical vapor deposition (CVD) were investigated by microscopy techniques. The CMSs were sufficiently mechanically elastic for extension
Journal of nanoscience and nanotechnology, 14(5), 3527-3531 (2014-04-17)
Pd-modified N-doped TiO2 nanoparticles were prepared by the sol-gel method. The X-ray diffraction (XRD) pattern indicated that the pure anatase TiO2 has been obtained. Transmission electron microscope (TEM) was used to observe the micro-morphology of the nanoparticles. The average size
The Journal of oral implantology, 40(2), 146-152 (2014-05-02)
Dental implants with an internal connection have been designed to establish a better stress distribution when lateral external forces act on the prosthesis and minimize the forces transmitted to the fastening screw. In the present study, 10 externally and 10
Journal of nanoscience and nanotechnology, 14(6), 4303-4307 (2014-04-18)
Na2Ti3O7 nanotubes was synthesized by a hydrothermal method, and characterized by X-ray diffraction (XRD), scanning electron microscopy (SEM), and transmission electron microscopy (TEM). Then the material was coated on Al2O3 ceramic substrate to fabricate humidity sensors using Ag-Pd as interdigitated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service