GF92630246
Praseodymium
foil, not light tested, 50x50mm, thickness 0.015mm, as rolled, 99%
Synonym(s):
Praseodymium, PR000150
About This Item
Assay
99%
form
foil
manufacturer/tradename
Goodfellow 926-302-46
resistivity
68 μΩ-cm, 20°C
bp
3520 °C (lit.)
mp
931 °C (lit.)
density
6.71 g/mL at 25 °C (lit.)
SMILES string
[Pr]
InChI
1S/Pr
InChI key
PUDIUYLPXJFUGB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | 1.03854 | 1.03859 | 85463 |
---|---|---|---|
according to ISO (10273 and FDA-BAM) | ISO 21872 | - | - |
Escherichia coli, nonselective for , coliforms | selective and differential for Vibrio spp. | nonselective for | nonselective and differential for Escherichia coli, nonselective and differential for Salmonella spp., nonselective and differential for coliforms, nonselective and differential for molds (General Media), nonselective and differential for yeasts (General Media) |
microbe id | utilization test: suitable, microbiological culture: suitable | microbiological culture: suitable | microbiological culture: suitable | microbe id | utilization test: suitable, microbiological culture: suitable |
food and beverages | food and beverages | food and beverages | agriculture |
200 | 200 | 200 | 100 |
General description
Legal Information
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service