Skip to Content
Merck
All Photos(2)

Key Documents

GF89275674

Indium

wire reel, 0.2m, diameter 1.0mm, as drawn, 99.999%

Synonym(s):

Indium, IN005130

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

≥99.999%

form

wire

manufacturer/tradename

Goodfellow 892-756-74

resistivity

8.37 μΩ-cm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The significance of chromosomal aberrations in indium-111-labeled lymphocytes.
M L Thakur et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 25(8), 922-927 (1984-08-01)
Indium-111 imaging in osteomyelitis and neuroarthropathy. Review and case report.
L M Techner et al.
Journal of the American Podiatric Medical Association, 76(1), 23-29 (1986-01-01)
[Scintigraphy with indium 111-labeled blood platelets].
C Kessler et al.
Deutsche medizinische Wochenschrift (1946), 109(48), 1853-1859 (1984-11-30)
[Use of 113m Indium as marker of compounds in the scintigraphy of the brain, lungs, liver, spleen, kidneys, placenta and heart].
A Mikolajków et al.
Polski tygodnik lekarski (Warsaw, Poland : 1960), 28(31), 1200-1201 (1973-07-30)
New generation of palladium-indium-silver dental cast alloys: a review.
M L Malhotra
Trends & techniques in the contemporary dental laboratory, 9(4), 65-68 (1992-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service