Skip to Content
Merck
All Photos(2)

Key Documents

GF74687080

Boron

monofilament, 10m, diameter 0.2mm

Synonym(s):

Boron, B 005915

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B
CAS Number:
Molecular Weight:
10.81
MDL number:
UNSPSC Code:
12141907
PubChem Substance ID:
NACRES:
NA.23

form

foil

manufacturer/tradename

Goodfellow 746-870-80

resistivity

1.5E12 μΩ-cm, 20 °C

L × diam.

10 m × 0.2 mm

density

2.34 g/mL at 25 °C (lit.)

SMILES string

[B]

InChI

1S/B

InChI key

ZOXJGFHDIHLPTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyoko Miwa et al.
Annals of botany, 105(7), 1103-1108 (2010-03-17)
The essentiality of boron (B) for plant growth was established > 85 years ago. In the last decade, it has been revealed that one of the physiological roles of B is cross-linking the pectic polysaccharide rhamnogalacturonan II in primary cell
Jan Fietzke et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(10), 2960-2965 (2015-02-26)
No records exist to evaluate long-term pH dynamics in high-latitude oceans, which have the greatest probability of rapid acidification from anthropogenic CO2 emissions. We reconstructed both seasonal variability and anthropogenic change in seawater pH and temperature by using laser ablation
Anne-Marie Alexander et al.
Chemical Society reviews, 39(11), 4388-4401 (2010-06-08)
Catalysts generated by the addition of carbon, nitrogen or phosphorus to transition metals have interesting properties and potential applications. The addition of carbon, nitrogen or phosphorus can lead to substantial modification of the catalytic efficacy of the parent metal and
Kyoko Miwa et al.
Advances in experimental medicine and biology, 679, 83-96 (2010-07-30)
Understanding of the molecular mechanisms of boron (B) transport has been greatly advanced in the last decade. BOR1, the first B transporter in living systems, was identified by forward genetics using Arabidopsis mutants. Genes similar to BOR1 have been reported
Boron containing compounds as protease inhibitors.
Reem Smoum et al.
Chemical reviews, 112(7), 4156-4220 (2012-04-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service