Skip to Content
Merck
All Photos(1)

Key Documents

GF58125966

Niobium-zirconium alloy, Nb99Zr1

200mm tube, 1.76mm outside diameter, 0.25mm wall thickness, 1.26mm inside diameter

Synonym(s):

NB037100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 581-259-66

L × wall thickness

200 mm × 0.25 mm

O.D. × I.D.

1.76 mm × 1.26 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service