GF51913942
Nickel/Cobalt
powder, Ni 97%/Co 3%, max. particle size 150 micron, weight 20 g, condition gas atomized
Synonym(s):
NE296010
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
powder
manufacturer/tradename
Goodfellow 519-139-42
weight
20 g
max. part. size
150 μm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service