GF42944744
Neodymium
rod, 2.0 mm diameter, length 50 mm, purity 99%
Synonym(s):
ND007902
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Nd
CAS Number:
Molecular Weight:
144.24
MDL number:
UNSPSC Code:
12141609
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99%
form
rod
manufacturer/tradename
Goodfellow 429-447-44
resistivity
64.0 μΩ-cm, 20°C
L
50 mm
diameter
2.0 mm
bp
3074 °C (lit.)
mp
1021 °C (lit.)
density
7.003 g/mL at 25 °C (lit.)
SMILES string
[Nd]
InChI
1S/Nd
InChI key
QEFYFXOXNSNQGX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service