Skip to Content
Merck
All Photos(2)

Key Documents

GF40007933

Tantalum

rod, 100mm, diameter 3.0mm, annealed, 99.9%

Synonym(s):

Tantalum, TA007926, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

99.9%

form

rod

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 400-079-33

resistivity

13.5 μΩ-cm, 20°C

L × diam.

100 mm × 3.0 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Tantalum dust as a contrast substance for laryngography and bronchography (a review of the literature)].
R M Rabinovich
Vestnik rentgenologii i radiologii, (1)(1), 77-79 (1981-01-01)
Ulrich Dorn et al.
The Journal of arthroplasty, 29(4), 831-835 (2013-10-09)
This study compared the corrosion behavior of tantalum-coated cobalt-chromium modular necks with that of titanium alloy modular necks at their junction to titanium-alloy femoral stem. Tests were performed in a dry assembly and two wet assemblies, one contaminated with calf
Xiao-kang Tang et al.
Zhongguo gu shang = China journal of orthopaedics and traumatology, 26(7), 617-620 (2013-10-19)
Incorrect treatment for femur head necrosis can cause collapse of femoral head and tresult in severe harm for the patients (especially for the patient with middle-aged and young). The structure and mechanics characteristics of tantalum rod is similar to bone
J Black
Clinical materials, 16(3), 167-173 (1993-12-09)
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and
Nilesh Patil et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 89(1), 242-251 (2008-10-08)
Conventional porous-coated joint prostheses used in hip and knee reconstruction have demonstrated good clinical results, however, these implants possess some inherent shortcomings such as low volumetric porosity, suboptimal frictional characteristics, and higher modulus of elasticity relative to that of bone.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service