GF36600599
Stainless Steel - AISI 304 alloy, FeCr18Ni10
foil, 100 x 100mm, 0.025mm thickness, hard, light tight (lt)
Synonym(s):
FE220221
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11171600
NACRES:
NA.23
Recommended Products
description
light tight (lt)
form
foil (hard
hard)
manufacturer/tradename
Goodfellow 366-005-99
size × thickness
100 mm × 100 mm × 0.025 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service