Skip to Content
Merck
All Photos(2)

Key Documents

GF28802695

Platinum

foil, 25x25mm, thickness 0.25mm, as rolled, 99.999%

Synonym(s):

Platinum, PT000271

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999%

form

foil

manufacturer/tradename

Goodfellow 288-026-95

resistivity

10.6 μΩ-cm, 20°C

size × thickness

25x25 mm × 0.25 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Medici et al.
Steroids, 66(2), 63-69 (2001-01-09)
Regioselectivity in the anodic electrochemical oxidation of cholic acid with different anodes is described. The oxidation with PbO(2) anode affords the dehydrocholic acid in quantitative yield after 22 h. 3alpha,12alpha-Dihydroxy-7-oxo-5beta-cholan-24-oic acid (59%) and 3alpha-hydroxy-7,12-dioxo-5beta-cholan-24-oic acid (51%) are obtained stopping the
Minimally invasive makeover: platinum foil technique.
Steve Ballis et al.
Dentistry today, 29(2), 146-146 (2010-03-04)
Yasar F Mansour et al.
Acta odontologica Scandinavica, 64(4), 231-236 (2006-07-11)
To quantify luting cement at the crown-tooth interface of esthetic crowns fabricated using four different techniques and two methods of excess cement removal. Four methods of crown fabrication were used: the feldspathic porcelain and platinum foil technique, the feldspathic porcelain
M Schuettler et al.
Journal of neural engineering, 2(1), S121-S128 (2005-05-07)
A new method for fabrication of microelectrode arrays comprised of traditional implant materials is presented. The main construction principle is the use of spun-on medical grade silicone rubber as insulating substrate material and platinum foil as conductor (tracks, pads and
Ayumi Ikeda et al.
Journal of medical and dental sciences, 52(4), 195-202 (2006-05-04)
The purpose of this study was to clarify the influence of changes of occlusal contacts on adjacent tooth displacement during articulation. The amount and direction of displacement of the maxillary left first molar and adjacent teeth were measured using a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service