Skip to Content
Merck
All Photos(2)

Key Documents

GF25800386

Vanadium

wire reel, 5m, diameter 0.5mm, as drawn, 99.8%

Synonym(s):

Vanadium, V 005120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

8 mmHg ( 20 °C)

Assay

99.80%

form

wire

manufacturer/tradename

Goodfellow 258-003-86

resistivity

24.8-26.0 μΩ-cm, 20°C

L × diam.

5 m × 0.5 mm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine Leblanc et al.
Biochimie, 88(11), 1773-1785 (2006-09-30)
Brown algal kelp species are the most efficient iodine accumulators among all living systems, with an average content of 1.0% of dry weight in Laminaria digitata, representing a ca. 30,000-fold accumulation of this element from seawater. Like other marine macroalgae
Anupam Bishayee et al.
Cancer letters, 294(1), 1-12 (2010-03-09)
Vanadium, a dietary micronutrient, is yet to be established as an essential part of the human diet. Over the past century, several biological effects of vanadium, such as insulin-mimetic action as well as amelioration of hyperlipidemia and hypertension, have been
Zu Liang Chen et al.
Analytica chimica acta, 607(1), 1-14 (2007-12-25)
A comprehensive review is presented addressing recent trends in the speciation and determination of vanadium in environmental and biological sample matrices, including important analytical aspects such as sample clean up, pre-concentration and method development. Methodology based on both separation and
D M Smith et al.
QJM : monthly journal of the Association of Physicians, 101(5), 351-358 (2008-03-06)
To assess the effectiveness of oral vanadium supplementation for glycaemic control in type 2 diabetes by conducting a systematic review of the literature. Eligible studies were identified by searching 14 databases using standardized terms. Experts, study authors and manufacturers were
Lise Coderre et al.
Canadian journal of physiology and pharmacology, 82(10), 833-839 (2004-12-02)
Inorganic and organic compounds of vanadium have been shown to exhibit a large range of insulinomimetic effects in the cardiovascular system, including stimulation of glucose transporter 4 (GLUT-4) translocation and glucose transport in adult cardiomyocytes. Furthermore, administration of vanadium compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service