Skip to Content
Merck
All Photos(2)

Key Documents

GF24172703

Beryllium

wire reel, 0.5m, diameter 0.25mm, annealed and clean, 99.7%

Synonym(s):

Beryllium, BE005115, Glucinium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Be
CAS Number:
Molecular Weight:
9.01
MDL number:
UNSPSC Code:
12141501
PubChem Substance ID:
NACRES:
NA.23

Assay

99.70%

form

wire

autoignition temp.

1198 °F

manufacturer/tradename

Goodfellow 241-727-03

resistivity

4.46 μΩ-cm, 20°C

L × diam.

0.5 m × 0.25 mm

bp

2970 °C (lit.)

mp

1278 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

SMILES string

[Be]

InChI

1S/Be

InChI key

ATBAMAFKBVZNFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elijah Thimsen et al.
Nature communications, 5, 5822-5822 (2014-12-20)
Thin films comprising semiconductor nanocrystals are emerging for applications in electronic and optoelectronic devices including light emitting diodes and solar cells. Achieving high charge carrier mobility in these films requires the identification and elimination of electronic traps on the nanocrystal
Michael J Brisson et al.
Journal of environmental monitoring : JEM, 8(6), 605-611 (2006-06-13)
Control of workplace exposure to beryllium is a growing issue in the United States and other nations. As the health risks associated with low-level exposure to beryllium are better understood, the need increases for improved analytical techniques both in the
L Mangin-Thro et al.
Nature communications, 6, 7705-7705 (2015-07-04)
The pseudo-gap phenomenon in copper oxide superconductors is central to any description of these materials as it prefigures the superconducting state itself. A magnetic intra-unit-cell order was found to occur just at the pseudo-gap temperature in four cuprate high-Tc superconducting
T Mark McCleskey et al.
Journal of occupational and environmental hygiene, 6(12), 751-757 (2009-11-07)
We compare beryllium to H+ and show that beryllium can displace H+ in many "strong hydrogen bonds" where Be as a "tetrahedral proton" (O-Be-O angle is tetrahedral as opposed to the nearly linear O-H-O angle) is thermodynamically preferred. The strong
D M Hollins et al.
Critical reviews in toxicology, 39 Suppl 1, 1-32 (2009-11-13)
The potential carcinogenicity of beryllium has been a topic of study since the mid-1940s. Since then, numerous scientific and regulatory bodies have assigned beryllium to various categories with respect to its carcinogenicity. Past epidemiologic and animal studies, however, have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service