GF04839767
Gold
tube, 100mm, outside diameter 1.60mm, inside diameter 0.6mm, wall thickness 0.5mm, as drawn, 99.95%
Synonym(s):
Gold, AU007180
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23
Assay
99.95%
form
tubes
manufacturer/tradename
Goodfellow 048-397-67
resistivity
2.05 μΩ-cm, 0°C
L × wall thickness
100 mm × 0.5 mm
O.D. × I.D.
1.60 mm × 0.6 mm
bp
2808 °C (lit.)
mp
1063 °C (lit.)
density
19.3 g/mL at 25 °C (lit.)
SMILES string
[Au]
InChI
1S/Au
InChI key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lin Luo et al.
Journal of nanoscience and nanotechnology, 14(6), 4036-4042 (2014-04-18)
A novel immunosensor of human immune globulin (IgG) was fabricated based on the fluorescence transfer between luminescent semiconductor quantum dots (QDs) and gold nanoparticles (AuNPs). AuNPs and CdSe/ZnS QDs were respectively labeled with immune reaction pair:IgG and goat anti-human immunoglobulin
Maria Jose Santos-Martinez et al.
Journal of biomedical nanotechnology, 10(6), 1004-1015 (2014-04-23)
The increasing use of gold nanoparticles in medical diagnosis and treatment has raised the concern over their blood compatibility. The interactions of nanoparticles with blood components may lead to platelet aggregation and endothelial dysfunction. Therefore, medical applications of gold nanoparticles
Jiajing Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4124-4138 (2014-04-18)
Gold nanostructures with promising applications in biomedical field have attracted great attention. However, some fundamental questions other than the development of novel applications should be elucidated before they can actually serve as biomedicines in the clinic. Bio-safety is one of
Cathleen M Crudden et al.
Nature chemistry, 6(5), 409-414 (2014-04-24)
Since the first report of thiol-based self-assembled monolayers (SAMs) 30 years ago, these structures have been examined in a huge variety of applications. The oxidative and thermal instabilities of these systems are widely known, however, and are an impediment to
Yingbo Zu et al.
Journal of biomedical nanotechnology, 10(6), 982-992 (2014-04-23)
Electroporation figured prominently as an effective nonviral gene delivery approach for its balance on the transfection efficiency and cell viability, no restrictions of probe or cell type, and operation simplicity. The commercial electroporation systems have been widely adopted in the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service