Skip to Content
Merck
All Photos(2)

Key Documents

G11608

Sigma-Aldrich

Guanidineacetic acid

99%

Synonym(s):

N-Amidinoglycine, N-Guanylglycine, Glycocyamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NHCH2CO2H
CAS Number:
Molecular Weight:
117.11
Beilstein:
1759179
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

300 °C (lit.)

SMILES string

NC(=N)NCC(O)=O

InChI

1S/C3H7N3O2/c4-3(5)6-1-2(7)8/h1H2,(H,7,8)(H4,4,5,6)

InChI key

BPMFZUMJYQTVII-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Stöckler et al.
Pediatric research, 36(3), 409-413 (1994-09-01)
In a patient with extrapyramidal movement disorder and extremely low creatinine concentrations in serum and urine, in vivo proton magnetic resonance spectroscopy disclosed a generalized depletion of creatinine in the brain. Oral substitution of arginine, a substrate for creatine synthesis
Joseph D T Ndika et al.
Molecular genetics and metabolism, 106(1), 48-54 (2012-03-06)
Arginine:glycineamidinotransferase (AGAT/GATM) deficiency has been described in 9 patients across 4 families. Here we describe the clinical outcome and response to creatine supplementation in a patient of the second family affected with AGAT deficiency-a 9-year-old girl. Delayed motor milestones were
Stefan Pilz et al.
Heart (British Cardiac Society), 97(15), 1222-1227 (2011-05-12)
Homoarginine is an amino acid which may play a role in nitric oxide and energy metabolism. Low homoarginine concentrations have previously been linked to increased mortality, but the underlying mechanisms remain unknown. To investigate whether serum homoarginine concentrations are associated
David Cheillan et al.
Orphanet journal of rare diseases, 7, 96-96 (2012-12-14)
A population of patients with unexplained neurological symptoms from six major French university hospitals was screened over a 28-month period for primary creatine disorder (PCD). Urine guanidinoacetate (GAA) and creatine:creatinine ratios were measured in a cohort of 6,353 subjects to
Saadet Mercimek-Mahmutoglu et al.
Molecular genetics and metabolism, 105(1), 155-158 (2011-10-25)
A 4-year-old female with history of developmental regression and autistic features was diagnosed with guanidinoacetate methyltransferase deficiency at age 21 months. Upon treatment, she showed improvements in her developmental milestones, sensorial-neural hearing loss and brain atrophy on cranial-MRI. The creatine/choline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service