Skip to Content
Merck
All Photos(1)

Key Documents

F17407

Sigma-Aldrich

1-Formylpiperidine

99%

Synonym(s):

N-Carbaldehyde piperidine, NFP, Piperidine-1-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
107697
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.1 mmHg ( 25 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.484 (lit.)

bp

222 °C (lit.)

density

1.019 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)N1CCCCC1

InChI

1S/C6H11NO/c8-6-7-4-2-1-3-5-7/h6H,1-5H2

InChI key

FEWLNYSYJNLUOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Direct amidation of azoles with formamides via metal-free C-H activation in the presence of tert-butyl perbenzoate
  • Vilsmeier-type reactions of pyrazoles with amides in the presence of phosphorous oxychloride
  • CO-free aminocarbonylation of N-substituted formamides with aryl iodides/bromides catalyzed by palladium acetate and Xantphos for synthesis of arylamides
  • Synthesis of alternating copolymers for organic photovoltaic applications
  • One-pot double functionalization of π-deficient heterocyclic lithium reagents
  • Synthesis of thermally stable piezofluorochromic aggregation-induced emission compounds

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hans Zahn et al.
Nature methods, 14(2), 167-173 (2017-01-10)
Single-cell genomics is critical for understanding cellular heterogeneity in cancer, but existing library preparation methods are expensive, require sample preamplification and introduce coverage bias. Here we describe direct library preparation (DLP), a robust, scalable, and high-fidelity method that uses nanoliter-volume
Janna Marie Bas-Hoogendam et al.
NeuroImage. Clinical, 26, 102247-102247 (2020-04-05)
Social anxiety disorder (SAD) runs in families, but the neurobiological pathways underlying the genetic susceptibility towards SAD are largely unknown. Here, we employed an endophenotype approach, and tested the hypothesis that amygdala hyperreactivity to faces conditioned with a social-evaluative meaning
Debajyoti Chatterjee et al.
Applied immunohistochemistry & molecular morphology : AIMM, 27(3), 210-215 (2017-08-12)
Pineal parenchymal tumor of intermediate differentiation (PPTID) is an uncommon tumor of the pineal gland. Although this behaves as a grade II/III tumor, the exact clinical behavior is not well known. There is no well-established pathologic factor that can predict
Nicolas Iván Bertone-Cueto et al.
Frontiers in systems neuroscience, 13, 78-78 (2020-01-31)
Field potentials (FPs) are easily reached signals that provide information about the brain's processing. However, FP should be interpreted cautiously since their biophysical bases are complex. The lateral habenula (LHb) is a brain structure involved in the encoding of aversive
Susan M Jack et al.
BMC health services research, 12, 50-50 (2012-03-02)
Despite an increase in knowledge about the epidemiology of intimate partner violence (IPV), much less is known about interventions to reduce IPV and its associated impairment. One program that holds promise in preventing IPV and improving outcomes for women exposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service