E8381
N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline sodium salt
Synonym(s):
DAOS
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
SMILES string
[Na].CCN(CC(O)CS(O)(=O)=O)c1cc(OC)cc(OC)c1
InChI
1S/C13H21NO6S/c1-4-14(8-11(15)9-21(16,17)18)10-5-12(19-2)7-13(6-10)20-3/h5-7,11,15H,4,8-9H2,1-3H3,(H,16,17,18)
InChI key
BTIDJAQNJLWPTI-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of diabetes investigation, 3(3), 235-244 (2012-06-06)
Aims/Introduction: High fat diet (HFD) is known to be a risk for development of type 2 diabetes. It is unclear, however, how it affects the glucose tolerance or the islet structure in type 2 diabetes. The aim of this study
AMB Express, 10(1), 176-176 (2020-10-03)
Biogenic amines (BAs) are low molecular weight organic bases formed by natural amino acids decarboxylation and trigger an array of toxicological effects in humans and animals. Bacterial amine oxidases enzymes are determined as practical tools to implement the rapid quantification
Bioscience, biotechnology, and biochemistry, 84(1), 118-125 (2019-09-14)
We developed an enzymatic assay system enabling easy quantification of 4-aminobutyric acid (GABA). The reaction of GABA aminotransferase obtained from Streptomyces decoyicus NBRC 13977 was combined to those of the previously developed glutamate assay system using glutamate oxidase and peroxidase.
The Analyst, 121(10), 1515-1518 (1996-10-01)
A spectrophotometric method was developed for the determination of ultratrace amounts of copper(II) based on its catalytic effect on the oxidative coupling reaction of 3-methyl-2-benzothiazolinone hydrazone with N-ethyl-N-(2-hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline to produce an intensely coloured dye (lambda(max) = 525 nm) in the
Enzymatic determination of serum total and esterified cholesterol with the same reagents.
Clinica chimica acta; international journal of clinical chemistry, 191(1-2), 97-99 (1990-10-31)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service