Skip to Content
Merck
All Photos(1)

Key Documents

DWKBWH0500PN

Azlon HDPE Wide Neck Round Bottles

screw cap, capacity 500 mL

Synonym(s):

BWH0500PN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43
capacity:
500 mL
material:
screw cap
wide-neck
packaging:
pkg of 10 ea

material

screw cap
wide-neck

packaging

pkg of 10 ea

manufacturer/tradename

Azlon BWH0500PN

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for a wide range of liquid storage and sampling applications

Features and Benefits

Suitable for food contact applications

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service