Skip to Content
Merck
All Photos(1)

Key Documents

D62209

Sigma-Aldrich

trans-1,2-Dichloroethylene

98%

Synonym(s):

trans-1,2-Dichloroethene, trans-Acetylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CHCl
CAS Number:
Molecular Weight:
96.94
Beilstein:
1420761
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5.16 psi ( 20 °C)

Quality Level

Assay

98%

form

liquid

expl. lim.

12.8 %

refractive index

n20/D 1.446 (lit.)

bp

48 °C (lit.)

mp

−50 °C (lit.)

density

1.257 g/mL at 25 °C (lit.)

SMILES string

Cl\C=C\Cl

InChI

1S/C2H2Cl2/c3-1-2-4/h1-2H/b2-1+

InChI key

KFUSEUYYWQURPO-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-1,2-Dichloroethylene is an organic solvent used in freon cleaning agents.

Application

trans-1,2-Dichloroethylene can be involved in the synthesis of:
  • (E)-Chloroenynes by reacting with 1-alkyne and piperidine in the presence of PdCl2(PPh3)2 catalyst in ethyl ether as a solvent.
  • (E)-3-trimethylsilyl-1-chloro-1-propene by reacting with trimethylsilylmethylmagnesium chloride (TMSCH2MgCl) in the presence of Co(II) or Co(III) acetylacetonate.
  • trans-1,2-Bis(tri-n-butylstannyl)ethylene which is an important reagent for the synthesis of myriad of natural products.
  • trans-1-Chloroalkenes by reacting with Grignard reagents in the presence of tetrakis(triphenylphosphine)nickel as catalyst.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Developmental toxicity of inhaled trans-1, 2-dichloroethylene in the rat
Hurtt, et al.
Fundamental and Applied Toxicology, 20, 225-230 (1993)
Suzuki Coupling Reactions of (E)-and (Z)-Chloroenynes with Boronic Acids: Versatile Access to Functionalized 1, 3-Enynes
Tikad A, et al.
European Journal of Organic Chemistry, 2010(4), 725-731 (2010)
Cobalt-catalyzed mono-coupling of R3SiCH2MgCl with 1, 2-dihalogenoethylene: a general route to γ-substituted (E)-allylsilanes.
Kamachi T, et al.
Tetrahedron Letters, 45(24), 4677-4679 (2004)
Preparation of trans-1, 2-bis(tributylstannyl) ethylene.
Corey EJ and Wollenberg RH
The Journal of Organic Chemistry, 40(25), 3788-3789 (1975)
A convenient preparation of trans(or cis)-1-chloroalkenes from trans(or cis)-1, 2-dichloroethylene: A new synthesis of the sex pheromone of lobesia botrana.
Ratovelomanana V and Linstrumelle G
Tetrahedron Letters, 22(4), 315-318 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service