Skip to Content
Merck
All Photos(1)

Key Documents

D4369

Sigma-Aldrich

2-Phenylacetophenone

97%

Synonym(s):

Deoxybenzoin, Benzyl phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2C6H5
CAS Number:
Molecular Weight:
196.24
Beilstein:
1072876
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

crystals

bp

320 °C (lit.)

mp

54-55 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C(Cc1ccccc1)c2ccccc2

InChI

1S/C14H12O/c15-14(13-9-5-2-6-10-13)11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

OTKCEEWUXHVZQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hadeel Elzeny et al.
Drug design, development and therapy, 11, 483-496 (2017-03-07)
Delivery of multiple therapeutics and/or diagnostic agents to diseased tissues is challenging and necessitates the development of multifunctional platforms. Among the various strategies for design of multifunctional nanocarriers, biodegradable polyphosphoester (PPE) polymers have been recently synthesized via a rapid and
Hassan Y Hijazi et al.
Journal of chromatography. A, 1617, 460824-460824 (2020-01-26)
A tailor-made porous molecularly imprinted polymer (MIP) thin-film was prepared by in situ photo-radical polymerization on a glass slide and used as a microextraction adsorbent. Detection was carried out using gas chromatography-mass spectrometry (GC-MS) to afford a method suitable for
Kwanghyok Jong et al.
Journal of biomaterials science. Polymer edition, 28(14), 1525-1537 (2017-05-24)
In this study, a novel type of pH-responsive polymer PyHES-NAC (2-hydroxy-3-(2-propynyloxy) propyl hydroxyethyl starch (PyHES)) - (N-acetyl-cysteine (NAC)) was synthesized. First, PyHES was prepared via hydrophobic modification of hydroxyl groups in hydroxyethyl starch (HES) with propynylglycidyl ether (PGE), and then
Zelal Adiguzel et al.
Materials science & engineering. C, Materials for biological applications, 78, 942-948 (2017-06-04)
This work describes a soft lithographic method for the generation of patterned both biopolymer and silver with each covered on microscope glass. Because of their biocompatible nature and permeability to gases the biopolymers are convenient for cell culture studies. The
Frederik Kotz et al.
Nature communications, 10(1), 1439-1439 (2019-03-31)
Fused silica glass is the preferred material for applications which require long-term chemical and mechanical stability as well as excellent optical properties. The manufacturing of complex hollow microstructures within transparent fused silica glass is of particular interest for, among others

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service