Skip to Content
Merck
All Photos(1)

Key Documents

D221953

Sigma-Aldrich

Dodecyl acetate

97%

Synonym(s):

Lauryl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2(CH2)11CH3
CAS Number:
Molecular Weight:
228.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

150 °C/15 mmHg (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCOC(C)=O

InChI

1S/C14H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-16-14(2)15/h3-13H2,1-2H3

InChI key

VZWGRQBCURJOMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth M MacDonald et al.
Journal of chemical ecology, 29(10), 2385-2389 (2003-12-20)
Larvae of the western flower thrips Frankliniella occidentalis are known to produce an anal droplet containing decyl acetate (10Ac) and dodecyl acetate (12Ac), which act as an alarm pheromone. Analysis by GC showed that the combined mass of 10Ac and
Developing new anti-cancer drugs.
Ricardo de Souza Pereira
Recent patents on anti-cancer drug discovery, 6(1), 1-5 (2010-11-30)
Ido Yosha et al.
Journal of agricultural and food chemistry, 56(17), 8045-8049 (2008-08-12)
Alginate-gelatin beads with dispersed droplets of a model pheromone, dodecyl acetate, were prepared as a vehicle for slow release of pheromones into the atmosphere over a prolonged period of time. The beads are prepared in two steps, the first being
K D Philipson et al.
The American journal of physiology, 248(1 Pt 2), H147-H150 (1985-01-01)
We have used charged amphiphiles as phospholipid analogues to modulate the interaction of Ca2+ with myocardial sarcolemma. The amphiphiles were dodecyl sulfate, dodecyltrimethylamine, and lauryl acetate; these are anionic, cationic, and neutral molecules, respectively. The hydrophobic alkyl chain is identical
B O Zimma et al.
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology, 189(10), 769-775 (2003-09-05)
The bumblebee Bombus (Psithyrus) norvegicus Sp.-Schn. is an obligate social parasite of B. (Pyrobombus) hypnorum L. Behavioural observations indicated that nest-invading B. norvegicus females may use allomones to defend themselves against attacking host workers. However, so far no defensive chemicals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service