Skip to Content
Merck
All Photos(1)

Key Documents

D139505

Sigma-Aldrich

4-(Dimethylamino)benzonitrile

98%

Synonym(s):

4-Cyano-N,N-dimethylaniline, DMABN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CN
CAS Number:
Molecular Weight:
146.19
Beilstein:
971606
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

bp

318 °C (lit.)

mp

72-75 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)C#N

InChI

1S/C9H10N2/c1-11(2)9-5-3-8(7-10)4-6-9/h3-6H,1-2H3

InChI key

JYMNQRQQBJIMCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Dimethylamino)benzonitrile is extensively used in photophysical studies due to its ability to undergo intramolecular charge transfer (ICT) from the dimethylamino moiety to the cyanophenyl moiety on photo-excitation leading to the appearance of dual fluorescence.

Application

4-(Dimethylamino)benzonitrile can be used in the synthesis of 3,6-diphenyl-2,5-dihydro-pyrrolo[3,4-c]pyrrole-1,4-dione derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The formation of a novel mercapturic acid during the metabolism of an N-methyl aromatic amine, 4-cyano-N,N-dimethylaniline.
C J Logan et al.
Biochemical pharmacology, 33(14), 2345-2346 (1984-07-15)
H S Rosenkranz et al.
Mutagenesis, 1(4), 275-282 (1986-07-01)
There was agreement between the experimental results, obtained in the course of the Second UKEMS Collaborative Study, for the mutagenicity in Salmonella typhimurium of benzidine, 4,4"-diaminoterphenyl, 4-dimethylaminoazobenzene and 4-cyanodimethylaniline and the mutagenicity predicted by CASE (Computer Automated Structure Evaluation), a
C J Logan et al.
Xenobiotica; the fate of foreign compounds in biological systems, 15(5), 391-397 (1985-05-01)
4-Cyano-N,N-dimethylaniline (CDA), when administered as a single oral dose to mice (18.5 mg/kg), was rapidly absorbed and eliminated. The major route of elimination was the urine (78% dose in 24h). The residues in the tissues 48 h after dosing, as
Fluorescence response of 4-(N, N?-Dimethylamino) benzonitrile in room temperature ionic liquids: observation of photobleaching under mild excitation condition and multiphoton confocal microscopic study of the fluorescence recovery dynamics.
Santhosh K, et al.
The Journal of Physical Chemistry B, 114(5), 1967-1974 (2010)
F K Ennever et al.
Mutagenesis, 1(2), 119-123 (1986-03-01)
The carcinogenicity prediction and battery selection (CPBS) procedure was used to analyze the short-term in vitro and in vivo genotoxicity results obtained during the Second UKEMS Collaborative Study. In accordance with preliminary animal bioassay results, CPBS predicted correctly the carcinogenicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service