Skip to Content
Merck
All Photos(1)

Documents

D12805

Sigma-Aldrich

3,5-Diaminobenzoic acid

98%, for peptide synthesis

Synonym(s):

3,5-DABA, 5-Carboxy-1,3-phenylenediamine, DABA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)2C6H3CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2086484
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

product name

3,5-Diaminobenzoic acid, 98%

Quality Level

Assay

98%

form

powder with small lumps

reaction suitability

reaction type: solution phase peptide synthesis

mp

235-238 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cc(N)cc(c1)C(O)=O

InChI

1S/C7H8N2O2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3H,8-9H2,(H,10,11)

InChI key

UENRXLSRMCSUSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Céline Douat-Casassus et al.
Journal of the American Chemical Society, 126(25), 7817-7826 (2004-06-24)
Peptide dendrimers incorporating 3,5-diaminobenzoic acid 1 as a branching unit (B) were prepared by solid-phase synthesis of ((Ac-A(3))(2)B-A(2))(2)B-Cys-A(1)-NH(2) followed by disulfide bridge formation. Twenty-one homo- and heterodimeric dendrimers were obtained by permutations of aspartate, histidine, and serine at positions A(1)
J Beckmann et al.
Acta diabetologica latina, 18(1), 51-57 (1981-01-01)
DNA content seems to be an ideal reference parameter for data on secretory function or metabolism of pancreatic islets. The approved fluorometric DNA assay with diaminobenzoic acid (DABA) of Kissane and Robins comprises repeated ethanol extractions of the tissue for
Interference of Bis-Tris buffer with the diaminobenzoic acid fluorescence assay used to quantify DNA.
W E Schy et al.
Mutation research, 226(4), 263-266 (1989-08-01)
[Improved fluorometric method for DNA microanalysis].
M M Vilenchik et al.
Izvestiia Akademii nauk SSSR. Seriia biologicheskaia, (2)(2), 217-222 (1984-03-01)
C F Kuo et al.
Analytical biochemistry, 170(1), 183-185 (1988-04-01)
A method for staining iron proteins on polyacrylamide gel electropherograms is described. It is based upon catalysis, by iron, of the oxidation of diaminobenzoic acid by hydrogen peroxide. Bands containing as little as 5 ng of protein-bound iron can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service