Skip to Content
Merck
All Photos(2)

Documents

D0791

Sigma-Aldrich

(Dimethylaminomethyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

impurities

~15% phenol
~20% bis(dimethylaminomethyl)phenol

SMILES string

CN(C)Cc1ccc(O)cc1

InChI

1S/C9H13NO/c1-10(2)7-8-3-5-9(11)6-4-8/h3-6,11H,7H2,1-2H3

InChI key

NFVPEIKDMMISQO-UHFFFAOYSA-N

Other Notes

Approx. 50% as mixture of o- and p-isomers

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service