CDS010483
(3R,3aS,6R,6aS,9S,10aS,10bR)-Octahydro-3,6,9-trimethyl-10aH-9,10b-epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one
AldrichCPR
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
SMILES string
C[C@@H]1CC[C@H]2[C@@H](C)C(=O)O[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23O4
InChI
1S/C15H22O4/c1-8-4-5-11-9(2)12(16)17-13-15(11)10(8)6-7-14(3,18-13)19-15/h8-11,13H,4-7H2,1-3H3/t8-,9-,10+,11+,13-,14-,15-/m1/s1
InChI key
ZQGMLVQZBIKKMP-NNWCWBAJSA-N
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Toxicology, 300(1-2), 57-66 (2012-06-09)
Artemisinin derivatives such as dihydroartemisinin (DHA) induce significant depletion of early embryonic erythroblasts in animal models. We have reported previously that DHA specifically targets pro-erythroblasts and basophilic erythroblasts, when human CD34+ stem cells are differentiated toward the erythroid lineage, indicating
Journal of natural products, 65(2), 184-188 (2002-02-23)
Photooxygenation of anhydrodeoxydihydroartemisinin (4) followed by chromatographic separation of the reaction mixture yielded the new compounds alpha- (5) and beta-hydroperoxydeoxyartemisitene (8) and the formate ester 7, together with two previously reported compounds, 6 and 9. Reduction of 5 using polymer-bound
Planta medica, 68(6), 515-518 (2002-07-03)
Artemisinin 1, dihydro-epideoxyarteannuin B 2 and deoxyartemisinin 3 were isolated from the sequiterpene lactone-enriched fraction obtained from the crude ethanolic extract of Artemisia annua L. These compounds were tested on ethanol and indomethacin-induced ulcer models. Compound 1 did not afford
Journal of medicinal chemistry, 38(26), 5038-5044 (1995-12-22)
A novel class of artemisinin analogs, N-alkyl-11-aza-9-desmethylartemisinins 17-29, were synthesized via ozonolysis and acid-catalyzed cyclization of precursor amides 5-16. These amides were prepared through condensation of an activated ester of the known intermediate acid 2 with the corresponding primary amine.
Journal of molecular graphics, 13(4), 215-222 (1995-08-01)
Artemisinin (qinghaosu, QHS) is a promising new antimalarial agent that is effective against drug-resistant strains of malaria. The antimalarial activity of this drug appears to be mediated by an interaction of the drug's endoperoxide bridge with intraparasitic hemin. We have
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service