Skip to Content
Merck
All Photos(1)

Key Documents

CDS000380

Sigma-Aldrich

1-Ethoxypropane

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O
CAS Number:
Molecular Weight:
88.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

CCCOCC

InChI

1S/C5H12O/c1-3-5-6-4-2/h3-5H2,1-2H3

InChI key

NVJUHMXYKCUMQA-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Puran Badkoobeh et al.
Iranian journal of reproductive medicine, 11(5), 355-364 (2014-03-19)
Doxorubicin (DOX), an anthracycline antibiotic, is a widely used anticancer agent. In spite of its high antitumor efficacy, the use of DOX in clinical chemotherapy is limited due to diverse toxicities, including gonadotoxicity. We investigated the protective effect of nano-zinc
Fadia S Youssef et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 23(12), 1484-1493 (2016-10-22)
The Australian plant Eremophila maculata F. Muell (Scrophulariaceae) is cultivated worldwide as an ornamental plant. This study was designed to assess the antioxidant and hepatoprotective activities of a methanol extract from E. maculata leaves (EMM) both in vitro and in
Vandana Kirar et al.
International journal of medicinal mushrooms, 19(11), 1009-1021 (2018-01-19)
Imbalanced oxygen availability is detrimental to normal cell function. Oxygen-sensitive cells such as cardiomyoblasts experience severe irreversible pathophysiological damage under conditions of reduced oxygen availability, such as hypoxia. A number of natural therapeutic agents have been explored for their potential
Lan Sun et al.
Molecules (Basel, Switzerland), 19(12), 19501-19515 (2014-11-29)
Platelet-derived growth factor-BB (PDGF-BB) and its downstream effector, extracellular signal-regulated kinase 1/2 (ERK1/2) MAP kinase, initiate a multitude of biological effects, including vascular smooth muscle cell (VSMC) proliferation and migration, which are critical events in the initiation and development of
Monique Michels et al.
Molecular medicine (Cambridge, Mass.), 21, 219-226 (2015-03-31)
Sepsis-associated encephalopathy (SAE) is associated with an increased rate of morbidity and mortality. It is not understood what the exact mechanism is for the brain dysfunction that occurs in septic patients, but brain inflammation and oxidative stress are a possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service