CADG5177
Dissolved gas analysis (DGA) glass syringe
capacity 100 mL, graduated, 4 mL, tip style, needle-lock Luer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
Recommended Products
feature
graduated 4 mL
tip style needle-lock Luer
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 5177
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
DGA glass syringes are used to take samples of mineral oil in a transformer. Transformer mineral oil cools the transformer and provides electrical insulation. When this mineral oil is subjected to high thermal and electrical stresses, it decomposes and gases are generated.
Different gases are generated from different types of faults, and the chemical analysis of these gases (Dissolved Gas Analysis) provides diagnostic information about the condition of the transformer.
DGA syringes are custom fit (matched numbered) to create a tight fit between the barrel and plunger. The barrels are frosted.
Different gases are generated from different types of faults, and the chemical analysis of these gases (Dissolved Gas Analysis) provides diagnostic information about the condition of the transformer.
DGA syringes are custom fit (matched numbered) to create a tight fit between the barrel and plunger. The barrels are frosted.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service