Skip to Content
Merck
All Photos(1)

Key Documents

C73400

Sigma-Aldrich

4-Methylbenzyl chloride

98%

Synonym(s):

α-Chloro-p-xylene, p-Xylyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2Cl
CAS Number:
Molecular Weight:
140.61
Beilstein:
507387
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

200 °C (lit.)

mp

4 °C (lit.)

density

1.062 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(CCl)cc1

InChI

1S/C8H9Cl/c1-7-2-4-8(6-9)5-3-7/h2-5H,6H2,1H3

InChI key

DMHZDOTYAVHSEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Małgorzata Pamuła et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(33), 7374-7383 (2020-02-23)
The synthesis of tetramethoxyresorcinarene podands bearing p-toluene arms connected by -SO3 - (1) and -CH2 O- (2) linkers is presented herein. In the solid state, the resorcinarene podand 1 forms an intramolecular self-inclusion complex with the pendant p-toluene group of
Electrophilic attack at carbon-5 of guanine nucleosides: structure and properties of the resulting guanidinoimidazole products.
R C Moschel et al.
IARC scientific publications, (70)(70), 235-240 (1986-01-01)
K Hemminki et al.
Journal of applied toxicology : JAT, 3(4), 203-207 (1983-08-01)
Benzyl chloride, benzyl bromide, p-methylbenzyl chloride, and p-nitrobenzyl chloride were used to study chemical reactivity with 4-(p-nitrobenzyl)-pyridine (NBP), and with guanosine in vitro, in relation to mutagenic potency in S. typhimurium and sister chromatid exchange (SCE) induction in CHO cells.
Nicholas R Oranzi et al.
Analytical chemistry, 91(6), 4092-4099 (2019-02-27)
Quantitation of the serum concentration of 25-hydroxyvitamin D is a high-demand assay that suffers from long chromatography time to separate 25-hydroxyvitamin D from its inactive epimer; however, ion mobility spectrometry can distinguish the epimer pair in under 30 ms due
Jae-Man Park et al.
Nature communications, 11(1), 4638-4638 (2020-09-17)
Existing gels are mostly polar, whose nature limits their role in soft devices. The intermolecular interactions of nonpolar polymer-liquid system are typically weak, which makes the gel brittle. Here we report highly soft and transparent nonpolar organogels. Even though their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service