Skip to Content
Merck
All Photos(3)

Key Documents

C109401

Sigma-Aldrich

Cyclooctane

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16
CAS Number:
Molecular Weight:
112.21
Beilstein:
1900349
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

16 mmHg ( 37.7 °C)

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.458 (lit.)

bp

151 °C/740 mmHg (lit.)

mp

10-13 °C (lit.)

density

0.834 g/mL at 25 °C (lit.)

SMILES string

C1CCCCCCC1

InChI

1S/C8H16/c1-2-4-6-8-7-5-3-1/h1-8H2

InChI key

WJTCGQSWYFHTAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclooctane can undergo oxidation with hydrogen peroxide in the presence of triethylpropylammonium-functionalized silica and transition metal mono-substituted polyoxotungstates.
Cyclooctane may undergo oxidation with hydrogen peroxide in the presence of triethylpropylammonium-functionalized silica and transition metal mono-substituted polyoxotungstates.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Silica supported transition metal substituted polyoxotungstates: Novel heterogeneous catalysts in oxidative transformations with hydrogen peroxide.
Estrada AC, et al.
Applied Catalysis A: General, 392(1-2), 28-35 (2011)
Heidi E Bostic et al.
Chemical communications (Cambridge, England), 48(10), 1431-1433 (2011-10-04)
Copper-free click chemistry was employed to derivatize membrane bilayers. This approach uses an azido-lipid conjugate presented on liposomes, which can be labeled in bioorthogonal fashion via cyclooctyne-tagged reagents. An immobilization-based approach using streptavidin-coated microplates was exploited to evaluate membrane derivatization.
Inigo Göttker-Schnetmann et al.
Journal of the American Chemical Society, 126(30), 9330-9338 (2004-07-30)
Reaction of bis(phosphinite) PCP iridium pincer complexes (p-XPCP)IrHCl (5a-f) [X = MeO (5a), Me (5b), H (5c), F (5d), C(6)F(5) (5e), Ar(F)(= 3,5-bis(trifluoromethyl)phenyl) (5f)] with NaOtBu in neat cyclooctane (COA) generates 1:1 mixtures of the respective (p-XPCP)IrH(2) complexes 4a-f and
The effect of a hydroxy protecting group on the PtCl2-catalyzed cyclization of dienynes--a novel, efficient, and selective synthesis of carbocycles.
Emily Mainetti et al.
Angewandte Chemie (International ed. in English), 41(12), 2132-2135 (2002-06-17)
Shawn Martin et al.
The Journal of chemical physics, 132(23), 234115-234115 (2010-06-25)
Understanding energy landscapes is a major challenge in chemistry and biology. Although a wide variety of methods have been invented and applied to this problem, very little is understood about the actual mathematical structures underlying such landscapes. Perhaps the most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service